ContactUsLinkComponent
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 5 Gal. Drawdown Tank, 5598406
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 5 Gal. Drawdown Tank, 5598406
Catalog # 76030-242
Supplier:  3M Healthcare
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 5 Gal. Drawdown Tank, 5598406
Catalog # 76030-242
Supplier:  3M Healthcare
Supplier Number:  7100079905
Specifications
  • Cat. No.:
    76030-242
  • Filter type:
    Tank
Specifications
About this item
3M™ Commercial reverse osmosis water storage tanks, model 5 gal. Drawdown tank is designed for use in steamer applications.
  • Reverse osmosis 5 gallon drawdown tank at 60 psi with 15 psi pre-charge
  • Includes connection kit