Order Entry
Puerto Rico
ContactUsLinkComponent
3M™ High Flow Series Twin Manifold Assembly (2XX), 6228505, 3M
3M™ High Flow Series Twin Manifold Assembly (2XX), 6228505, 3M
Catalog # 76028-736
Supplier:  3M Healthcare
undefined
3M™ High Flow Series Twin Manifold Assembly (2XX), 6228505, 3M
Catalog # 76028-736
Supplier:  3M Healthcare
Supplier Number:  7000001833

Specifications

  • Cat. No.:
    76028-736
  • Pressure range:
    172 - 862 kPa (25 - 125 psig)
  • Filter type:
    SQC filtration system
  • Inlet size:
    ¾" MNPT
  • Max. operating temperature:
    37.8 °C (100 °F)

Specifications

About this item

3M™ High Flow Series Twin Manifold Assembly (2XX) includes a single inlet port for incoming water and a single outlet port for filtered water.

  • ¾" MNPT inlet and outlet water connections
  • Parallel flow
  • Designed for use with 3M™ High Flow Series water filtration cartridges
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench

Filter manifold assemblies provide a vehicle for appropriate water quality to equipment requiring different water characteristics for various applications. These products are innovatively designed for ease of installation and maintenance. Most manifold models include a pressure gauge to monitor differential water pressure.