- Cat. No.:76030-531
- Model:5628103
3M™ Water Filtration Products OCS In-line Systems for Office Water Applications provide you with consistent quality water for bottle-less water coolers and cold water dispensers.
- Provides consistent quality water for bottle-less water coolers and cold water dispensers by reducing sediment* and chlorine taste and odor
- Reduction of sediment helps prevent equipment wear and can help extend the life of o-rings and seals
- Encapsulated cartridge design allows for fast and easy cartridge change-outs
- Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
- Compact design allows for installation in tight spaces
Our 3M™ Water Filtration Products OCS In-line systems for Office Water Applications are an inexpensive way to help protect your equipment while providing cleaner, clearer, better tasting water
The Sanitary Quick Change (SQC) design allows for fast and easy cartridge change-outs that are sanitary by design – requiring no contact with the filter media during cartridge change-out.
3M™ Water Filtration Products branded replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
*Tested and verified by manufacturer’s laboratory.