Specifications
- Cat. No.:76030-529
- Model:5626210
Specifications
About this item
3M™ Water Filtration Products OCS System for Office Coffee Applications featuring VH3 Valve-in-Head & Scale Reduction provides you with consistent quality water for hot beverages that can help increase repeat business and help to protect your equipment from hard scale build-up.
- Provides consistent quality water for office coffee and hot cup applications by reducing sediment,* scale and chlorine taste and odor
- Reduces the ability of calcium and magnesium to form as hard scale* helping to reduce downtime and unscheduled service maintenance
- Reduction of sediment* can help prevent equipment wear
- Built-in valve allows cartridge change-outs without shutting off the incoming water supply, no external shut-off valve required
- Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
Our 3M™ Water Filtration Products OCS System for Office Coffee Applications featuring VH3 Valve-in-Head & Scale Reduction for brewed coffee, tea and hot chocolate is designed to be effective at reducing sediment*, chlorine taste and odor, and scale* allowing you to feel more confident in the quality of the products you serve and can provide equipment protection from scale build-up
The Sanitary Quick Change (SQC) design allows for fast and easy cartridge change-outs and are sanitary by design, requiring no contact with the filter media during cartridge change-out. The ramped interconnect completely disconnects the cartridge from the filter head with only a /14; turn, making this an extremely easy cartridge change-out.
3M™ Water Filtration Products replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
*Tested and verified by manufacturer's laboratory.