Specifications
- Ø:113.3 mm (4.46")
- Flow rate:1.9 lpm (0.5 gpm)
- Micron Rating:1.0
- Capacity:4727 L (1249 gal.)
- Length:526.3 mm (20.72")
- Cat. No.:76031-718
- Pressure range:172 - 862 kPa (25 - 125 psig)
- Filter type:SQC filtration system
- Model:5630501
- Max. operating temperature:37.8 °C (100 °F)
Specifications
About this item
3M™ ScaleGard™ blend series of cartridges for hardness and chloramines reduction delivers custom water quality that you control. Simply blend in the desired level of minerals for Recipe Quality Water™. Cartridges reduce chlorine taste and odor – as well as chloramines that can react with metal, plastic and rubber materials; this reaction may potentially cause equipment damage.
- Easy-to-operate adjustable dial for precision blending when using 3M™ ScaleGard™ blend series of heads
- Premium carbon block technology provides consistent Recipe Quality Water™ for coffee, espresso, vending, steamers and combination ovens
- High-capacity buffered ion exchange resin reduces calcium and magnesium ions that can cause hard scale build-up on equipment
- Sanitary quick change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
- Helps reduce corrosion potentially caused by chloramines1
- Manufactured from FDA CFR-21 compliant materials*
- Reduce water hardness and help maintain equipment efficiency
Our 3M™ ScaleGard™ blend series quadruple-action filtration cartridges provide protection against hard scale, corrosion, chlorine taste and odor; and chloramines. Reduce water hardness and help maintain equipment efficiency with our 3M™ ScaleGard™ Blend Series of Cartridges for Hardness and Chloramines Reduction. These high-capacity ion exchange systems will lessen bitter chlorine and chloramines tastes while helping to prevent scale build-up to help you provide great tasting beverages.
In the competitive foodservice industry, product quality and consistency are critical to business success. Equipment downtime and unscheduled service calls threaten customer satisfaction and profit margins. It's very important to have competitive and functional scale reduction that helps protect equipment and leaves the right elements in the water. Scale reduction will allow you to make great tasting hot beverages.
Our global tradition of innovation in filtration technology
Having cleaner and clearer water for your business is both a pleasure and a reassurance that you're giving your customers Recipe Quality Water™. With a water filtration system from 3M purification, you can provide filtered water that will help protect your equipment and your profits. From commercial point-of-entry water filtration systems to dedicated point-of-use water filtration systems, look to 3M purification to help you find the right solution.
For more than a century, 3M has applied innovation and technology to improve our customers' lives and support their business goals. From our headquarters in St. Paul, Minnesota, we serve industrial and commercial customers and consumers in nearly 200 countries. Across thousands of products, the 3M brand represents consistency, superior quality and value. Our success begins with a culture of creative collaboration that inspires a never ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies - often in combination - to an endless array of real-world customer needs. Of course, all of this is made possible by the people of 3M and our singular commitment to make life easier and better for people around the world. Toward that end, 3M is committed to providing innovative, cost-effective products that help protect people, equipment and our environment. 3M Purification Inc. (formerly CUNO Incorporated), provides filtration and separation solutions for residential, food service, science labs, industrial, marine and other applications.
*Surfaces that are in contact with water are FDA CR-21 compliant.
1Tested and verified by manufacturer's laboratory.