ContactUsLinkComponent
3M™ ScaleGard™ Commercial Reverse Osmosis System for Boilerless Steamers and Combi-Ovens, Model SGLP100-CL, 5636201
3M™ ScaleGard™ Commercial Reverse Osmosis System for Boilerless Steamers and Combi-Ovens, Model SGLP100-CL, 5636201
Catalog # 76028-714
Supplier:  3M Healthcare
3M™ ScaleGard™ Commercial Reverse Osmosis System for Boilerless Steamers and Combi-Ovens, Model SGLP100-CL, 5636201
Catalog # 76028-714
Supplier:  3M Healthcare
Supplier Number:  7100093217
Specifications
  • Cat. No.:
    76028-714
  • Model:
    5636201
Specifications
About this item
3M™ ScaleGard™ Commercial Reverse Osmosis System for Boilerless Steamers and Combi-Ovens, Model SGLP100-CL featuring scale and chloramines reduction are designed to reduce hardness minerals, sediment, chloramines and chlorides that can cause equipment downtime and decrease profitability.
  • Helps reduce corrosion potentially caused by chlorides and chloramines
  • Reduces high TDS and high hardness to produce Recipe Quality Water™ for boilerless steamer and combi-oven applications
  • Helps to reduce scale build-up that can make cleaning difficult in steamers
  • Reduced equipment maintenance caused by lime scale build-up
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
In the competitive foodservice industry, product quality and consistency are critical to business success
Equipment downtime and unscheduled service calls threaten customer satisfaction and profit margins. Our 3M™ Water Filtration Products SGLP-CL Series Systems are designed to help maintain efficient equipment operation in water conditions with high TDS and/or high hardness. Our uniquely designed 3M™ ScaleGard™ Commercial Reverse Osmosis System for Boilerless Steamers and Combi-Ovens, Model SGLP100-CL not only help reduce Hardness and TDS; they also combat the effects of Chlorides and Chloramines, which can create a corrosive environment that can potentially lead to premature equipment failure.
When water is cooled, heated or converted to steam, it leaves impurities behind, such as hardness minerals. These impurities cling to solid surfaces and can, over time, build up and cause problems. Our 3M™ SGLP-CL Series Systems are designed to reduce hardness minerals, sediment, chloramines and chlorides that can cause equipment downtime and decrease profitability.