Specifications
- Ø:113.3 mm (4.46")
- Flow rate:1.9 lpm (0.5 gpm)
- Micron Rating:1.0
- Capacity:1604 L (423 gal.)
- Length:297.7 mm (11.72")
- Cat. No.:76031-720
- Pressure range:172 - 862 kPa (25 - 125 psig)
- Filter type:SQC filtration system
- Model:5630801
- Max. operating temperature:37.8 °C (100 °F)
Specifications
About this item
3M™ ScaleGard™ Pro series of cartridges for hardness and chloramines reduction reduce chlorine taste and odor – as well as chloramines that can react with metal, plastic and rubber materials; this reaction may cause equipment damage. Our high-capacity ion exchange scale reduction media helps provide superior equipment protection from hard scale and corrosion.
- Premium carbon block technology provides consistent Recipe Quality Water™ for coffee, espresso, vending, steamers and combination ovens
- High-capacity buffered ion exchange resin reduces calcium and magnesium ions that can cause hard scale build-up on equipment
- Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
- Helps reduce corrosion potentially caused by Chloramines1
- Manufactured from FDA CFR-21 compliant materials*
Reduce water hardness and increase equipment efficiency 3M™ ScaleGard™ Pro series quadruple-action filtration cartridges provide protection against hard scale, corrosion, chlorine taste and odor and chloramines.
Reduce water hardness and increase equipment efficiency with our 3M™ ScaleGard™ Pro series of cartridges for hardness and chloramines reduction. Our ion exchange systems will lessen bitter chlorine and chloramines tastes while decreasing scale build-up to ensure you produce the best tasting beverages possible.
In the competitive foodservice industry, product quality and consistency are critical to business success. Equipment downtime and unscheduled service calls threaten customer satisfaction and profit margins. The need for competitive and functional scale reduction that protects equipment and leaves the right elements in the water to make great tasting hot beverages has never been more important.
3M water filtration products branded replacement cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided by many 3M water filtration products.
Our global tradition of innovation in filtration technology for more than a century, 3M has applied innovation and technology to improve our customers' lives and support their business goals. From our headquarters in St. Paul, Minnesota, we serve industrial and commercial customers and consumers in nearly 200 countries. Across thousands of products, the 3M brand represents consistency, superior quality and value. Our success begins with a culture of creative collaboration that inspires a never ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies - often in combination - to an endless array of real-world customer needs. Of course, all of this is made possible by the people of 3M and our singular commitment to make life easier and better for people around the world. Toward that end, 3M is committed to providing innovative, cost-effective products that help protect people, equipment and our environment. 3M Purification Inc. (formerly CUNO Incorporated), provides filtration and separation solutions for residential, food service, science labs, industrial, marine and other applications.
1Tested and verified by manufacturer's laboratory.