ContactUsLinkComponent
3M™ Valve-In-Head/Bracket Assembly, Model VH3-NPT, 6213033, 3M
3M™ Valve-In-Head/Bracket Assembly, Model VH3-NPT, 6213033, 3M
Catalog # 76030-649
Supplier:  3M Healthcare
3M™ Valve-In-Head/Bracket Assembly, Model VH3-NPT, 6213033, 3M
Catalog # 76030-649
Supplier:  3M Healthcare
Supplier Number:  7100163109
Specifications
  • Cat. No.:
    76030-649
Specifications
About this item
3M™ Water Filtration Products High Flow Series NEP Valve-In-Head (VH3) with shroud and gauge.
  • ⅜" FNPT, Valve-in-head with Shroud and Gauge
  • FDA CFR-21 compliant materials
  • NSF Certified