Order Entry
Puerto Rico
ContactUsLinkComponent
HIV TAT-GluR23A Fusion Peptide
HIV TAT-GluR23A Fusion Peptide
Catalog # 103008-148
Supplier:  Anaspec Inc
CAS Number:  
HIV TAT-GluR23A Fusion Peptide
Catalog # 103008-148
Supplier:  Anaspec Inc
Supplier Number:  AS-64984
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    HIV
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    TAT fusion|Fusion TAT GluR23Y|GluR23Y TAT
  • Protein/Peptide Name:
    TAT-GluR23A Fusion Peptide
  • Purity:
    95%
  • Molecular Weight:
    2357.7 Da
  • Sequence:
    YGRKKRRQRRRAKEGANVAG
  • Cat. No.:
    103008-148
  • Supplier no.:
    AS-64984

Specifications

About this item

This is the GluR23A sequence, a control inactive peptide used as a mutant counterpart to glutamate receptor endocytosis inhibitor (GluR23Y), connected to an 11 amino acid cell permeable HIV Trans-Activator of Transcription (TAT) protein transduction domain (PTD). GluR23A is derived from GluR23Y amino acids 869 to 877, with Ala substituted for Tyr, and thus lacking essential phosphorylation sites.
Sequence: YGRKKRRQRRRAKEGANVAG
MW: 2357.7 Da
% Peak area by HPLC: 95
Storage condition: -20°C