Order Entry
Puerto Rico
ContactUsLinkComponent
Human ACE Substrate 2
Human ACE Substrate 2
Catalog # 103005-964
Supplier:  Anaspec Inc
CAS Number:  
Human ACE Substrate 2
Catalog # 103005-964
Supplier:  Anaspec Inc
Supplier Number:  AS-60758
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    ACE Caspase I substrate|Angiotensin I Converting Enzyme 2 Substrate|ACE substrate|ACE substrate 2|ACE 2 Caspase substrate
  • Protein/Peptide Name:
    ACE Substrate 2
  • Purity:
    95%
  • Molecular Weight:
    1145.2 Da
  • Sequence:
    Mca-YVADAPK(Dnp)
  • Cat. No.:
    103005-964
  • Supplier no.:
    AS-60758

Specifications

About this item

Cleavage of this FRET substrate generates the fluorescent Mca signal that is monitored fluorimetrically at 393 nm, with excitation at 325 nm. A highly fluorescent substrate for caspase 1.
Sequence: Mca-YVADAPK(Dnp)
MW: 1145.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C