Order Entry
Puerto Rico
ContactUsLinkComponent
EBV (Epstein-Barr virus) CEF EBV
EBV (Epstein-Barr virus) CEF EBV
Catalog # 102997-156
Supplier:  Anaspec Inc
CAS Number:  
EBV (Epstein-Barr virus) CEF EBV
Catalog # 102997-156
Supplier:  Anaspec Inc
Supplier Number:  AS-28319
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    EBV (Epstein-Barr virus)
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    CEF10 EBV (426-434)|EBV CEF10|CEF LMP2
  • Protein/Peptide Name:
    CEF EBV
  • Purity:
    95%
  • Molecular Weight:
    906.2 Da
  • Sequence:
    CLGGLLTMV
  • Cat. No.:
    102997-156
  • Supplier no.:
    AS-28319

Specifications

About this item

HLA A2.1-restricted epitope from Epstein-Barr Virus latent membrane protein LMP2 (426-434).
Sequence: CLGGLLTMV
MW: 906.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C