Order Entry
Puerto Rico
ContactUsLinkComponent
Influenza CEF Influenza
Influenza CEF Influenza
Catalog # 102997-138
Supplier:  Anaspec Inc
CAS Number:  
Influenza CEF Influenza
Catalog # 102997-138
Supplier:  Anaspec Inc
Supplier Number:  AS-28310
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Influenza
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    CEF M1 Protein|CEF1 Influenza (58-66)|Influenza Matrix CEF1
  • Protein/Peptide Name:
    CEF Influenza
  • Purity:
    95%
  • Molecular Weight:
    966.2 Da
  • Sequence:
    GILGFVFTL
  • Cat. No.:
    102997-138
  • Supplier no.:
    AS-28310

Specifications

About this item

HLA A2-restricted epitope from influenza matrix protein (58-66).
Sequence: GILGFVFTL
MW: 966.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C