Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:CEF Peptide|CEF Control Pool|CEF Pool
- Protein/Peptide Name:CEF Control Peptide Pool
- Purity:95%
- Tested Applications:Control Peptide
- Cat. No.:103006-274
- Supplier no.:AS-61036-003
This product contains 0.03 mg (net) of the 32 CEF peptides for a total of 1 mg in one vial. Used in the stimulation of IFNγ release from CD8+ T cells in individuals with defined HLA types, these epitopes are useful in applications such as ELISPOT, intracellular cytokine and CTL assays.
% peak area by HPLC: ≥ 95
Storage condition: -20°C