Order Entry
Puerto Rico
ContactUsLinkComponent
Chicken OVA-G4 Peptide
Chicken OVA-G4 Peptide
Catalog # 103008-044
Supplier:  Anaspec Inc
CAS Number:  
Chicken OVA-G4 Peptide
Catalog # 103008-044
Supplier:  Anaspec Inc
Supplier Number:  AS-64384
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Chicken
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    OVA-G4|SIIGFEKL Peptide|G4 Peptide|OVA variant Peptide
  • Protein/Peptide Name:
    OVA-G4 Peptide
  • Purity:
    95%
  • Molecular Weight:
    906.1 Da
  • Sequence:
    SIIGFEKL
  • Cat. No.:
    103008-044
  • Supplier no.:
    AS-64384

Specifications

About this item

G4 peptide (SIIGFEKL) is a variant of the agonist ovalbumin (OVA) peptide (257-264), SIINFEKL. OVA Peptide is a class I (Kb)-restricted peptide epitope of ovalbumin presented by the class I MHC (major histocompatibility complex) molecule, H-2Kb (class I genes of the mouse MHC).
Sequence: SIIGFEKL
MW: 906.1 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C