Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Species:Human
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:MOG T cells|pMOG|MOG-specific T-cells|MOG CD8+|MOG-specific CD8+ T cells|MOG(44-55)
- Protein/Peptide Name:Peptide myelin oligodendrocyte glycoprotein
- Purity:95%
- Molecular Weight:1347.6 Da
- Sequence:FSRVVHLYRNG
- Cat. No.:103006-880
- Supplier no.:AS-61620
Specifications
About this item
This 11 mer peptide myelin oligodendrocyte glycoprotein ( pMOG) ( 44–54) represents the core binding epitope for MOG-specific CD8+ T cells. This is the minimal, functionally constant, length of the MOG (35–55) peptide that binds to CD8+ MOG-specific T cells and stimulates T cell proliferation in vitro.
Sequence: FSRVVHLYRNG
MW: 1347.6 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C