Order Entry
Puerto Rico
ContactUsLinkComponent
TraceSELECT™ Fluka™ Cesium Chloride - Lanthanum Chloride Buffer, Honeywell Reseach Chemicals
TraceSELECT™ Fluka™ Cesium Chloride - Lanthanum Chloride Buffer, Honeywell Reseach Chemicals
Catalog # BJ95164-250ML
CAS Number:  
TraceSELECT™ Fluka™ Cesium Chloride - Lanthanum Chloride Buffer, Honeywell Reseach Chemicals
Catalog # BJ95164-250ML
Supplier Number:  95164-250ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle
  • Description:
    Cesium chloride Lanthanum chloride buffer
  • Size:
    250 ml
  • Cat. No.:
    BJ95164-250ML

Specifications

About this item

TraceSELECT™, for flame atomic absorption spectrometry.

  • Packaged in high-quality containers appropriate for each product
  • The reagents are produced and bottled under clean-room conditionsions

Lanthanum chloride (LaCl3) 99.0 - 101.0 g/l
Cesium chloride (Cscl) 99.0 - 101.0 g/l
Density (D 20/4) 1.152 - 1.176
Calcium (Ca) max. 0.5 ppm