HistoPro 200 Parafin Infiltration Processor, Electron Microscopy Sciences
Product Details & Documents
HistoPro® 200 offers unique reagent/paraffin infiltration capability with programmable processing time, temperature, agitation and vacuum in an easy to use, compact, cost conscious package.
- Up to 100 Cassettes
- 2 Liter Reagent/Paraffin Capacity
- Compact footprint
- Programmable processing time and temperature
- Programmable vacuum cycle
Certifications: UL listed and CE certified.
Specifications
- Power:150 W
- Description:HistoPro 200 Parafin Infiltration Processor
- W×D×H:23 x 51 x 38 cm (9 x 20 x 38" )
- Electrical:100/230 VAC, 50/60 Hz
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



