Pierce™ Protein A Agarose, Thermo Scientific
Supplier: Invitrogen
Certificates
About this item
Protein A Agarose is a standard-capacity Protein A beaded agarose resin for use in a variety of antibody affinity purification methods.
- Immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
- Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
- Protein A is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non-specific binding and enabling multiple uses without decline in yield
- Standard capacity beads with a normal load of immobilized Protein A
Specifications
- Binding Capacity:12 – 19 mg Human IgG/mL