Order Entry
Puerto Rico
ContactUsLinkComponent
 
Single FIB Sample and Grid Holder, Electron Microscopy Sciences
  103306-232
Single FIB Sample and Grid Holder, Electron Microscopy Sciences
  103306-232
 :  75950-03

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Single FIB Sample and Grid Holder, Pin
  • Dimensions:
    1/2" (12.7 mm)
  • Cat. No.:
    103306-232
  • Supplier no.:
    75950-03

 

 

Single FIB sample and grid holder holds a FIB sample mounted on standard 1/2" (12.7 mm) pin stub for FIB milling and lift-out procedures.

  • Vacuum grade aluminum with brass screws
  • Holds two FIB grids close to the sample to mount prepared TEM lamellae on the FIB grid for TEM imaging
  • Suitable for all FIB/SEM systems which accept pin mount holders, including the FEI, ZEISS and Tescan systems
  • Cost-effective holder

Overall dimensions: 22.4 x 12.7 x 11.7 mm (0.88" x 0.5" x 0.46")
Pin: Standard 3.2 mm (1/8")

 : Includes Philips screwdriver.