Order Entry
Puerto Rico
ContactUsLinkComponent
Human Beta-Amyloid (1-15)-Lys16, HiLyte™ Fluor 488
Human Beta-Amyloid (1-15)-Lys16, HiLyte™ Fluor 488
Catalog # 103007-136
Supplier:  Anaspec Inc
CAS Number:  
Human Beta-Amyloid (1-15)-Lys16, HiLyte™ Fluor 488
Catalog # 103007-136
Supplier:  Anaspec Inc
Supplier Number:  AS-62002-05
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    HiLyte™ Fluor 488
  • Species:
    Human
  • Size:
    0.5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    beta amyloid 1-16 Lys HiLyte™ Fluor 488|amyloid 1-16 Lys HiLyte™ Fluor 488|abeta (1-16) Lys HiLyte™ Fluor 488|amyloid beta (1-16) Lys HiLyte™ Fluor 488
  • Protein/Peptide Name:
    beta-Amyloid (1-15)-Lys16
  • Purity:
    95%
  • Molecular Weight:
    2311.4 Da
  • Sequence:
    DAEFRHDSGYEVHHQ-K(HiLyte™ Fluor 488)
  • Cat. No.:
    103007-136
  • Supplier no.:
    AS-62002-05

Specifications

About this item

This peptide is the beta-Amyloid peptide, residues 1 to 16 labeled with HiLyte™ Fluor 488 on the Lys16, Abs/Em =501/527.
Sequence: DAEFRHDSGYEVHHQ-K(HiLyte™ Fluor 488)
Molecular Weight: 2311.4 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C