About this item
Aß (12–28) residues are the binding site for apolipoprotein E (apoE) on Aß. This sequence encompasses a hydrophobic domain (residues 14–21) and a ß-turn (residues 22–28) which place two hydrophobic domains of Aß 14 to 21 and 29 to 40/42 opposite each other, allowing for the assembly of Aß peptides into fibrils. The secondary structure of Aß (12- 28), a neutral peptide, is dominated by a-helix and random coil.
equence: VHHQKLVFFAEDVGSNK
Molecular Weight: 1955.2 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Protein Synonyms:amyloid beta (12-28)|abeta (12-28)|beta amyloid 12-28|amyloid 12-28
- Protein/Peptide Name:beta-Amyloid (12-28)
- Purity:95%
- Molecular Weight:1955.2 Da
- Sequence:VHHQKLVFFAEDVGSNK