ContactUsLinkComponent
Cardiac Microvascular Endothelial Cells, Lifeline Cell Technology
Cardiac Microvascular Endothelial Cells, Lifeline Cell Technology
Catalog # 76035-952
CAS Number:  
undefined
Cardiac Microvascular Endothelial Cells, Lifeline Cell Technology
Catalog # 76035-952
Supplier Number:  FC-0053
CAS Number:  
Specifications
  • Cell Format:
    Cryopreserved
  • Single Donor/Pooled:
    Pooled
  • Cat. No.:
    76035-952
Specifications
About this item
Lifeline® Normal Human Cardiac Microvascular Endothelial Cells, when grown in Lifeline® VascuLife® VEGF-Mv Medium, provide an ideal low-serum culture model, for the study of angiogenesis, atherosclerosis, or vascular biology.
Isolated from human heart tissue
Quaity tested for growth and morphology over at least 15 population doublings
Unexposed to antimicrobials or phenol red
Lifeline® Human Cardiac Microvascular Endothelial Cells are isolated from human heart tissue, plated onto culture vessels, and expanded in culture vessels three times before being harvested for cryopreservation.  
Cardiac Microvascular Endothelial Cells are quality tested to ensure optimal growth and morphology over a period of at least 15 population doublings.
Lifeline® Human Microvascular Endothelial Cells are not exposed to antimicrobials or phenol red when cultured in VascuLife® Media.