Microscope Slide Storage Box with Removable Tray, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
The rugged and durable microscope slide storage boxes feature a hinged lid for protection while providing easy viewing and access to your microscope slides.
- Transparent lid provides clear view of contents without opening
- Removable draining tray holds 100 slides in pre-numbered slots
- Tray provides space between each slide so slides can be tilted forward or back for easy identification
The trays can accommodate up to 200 standard sized microscope slides (2 I each pre-numbered slot) on end for space savings.
Ordering information: Removable draining trays are also available separately.