Methionine Sulfoxide Immunoblotting Kit, Cayman Chemical
Catalog # 75816-974
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Brand Name:CAYM
- Cat. No.:75816-974
- Contents:Kit contains: Antibody, Positive Control, and Blocking Reagent
- Supplier no.:600160-1
Specifications
About this item
For the immunochemical detection of proteins containing MetO residues by western blotting.
- Detect proteins containing MetO residues by Western blot
- Positive control included
Protein MetO is a reversible oxidative modification that occurs by exposure of protein(s) methionine residues to reactive oxygen species (ROS). Methionine oxidation can alter the function(s) of the modified proteins and if not reversed by MetO reductases can be further oxidized to methionine sulfone, an irreversible modification. Cayman’s MetO Immunoblotting Kit contains reagents needed for the immunochemical detection of proteins containing MetO residues by western blotting. MetO-containing samples of interest include those from cell or tissue lysates as well as semi-pure or purified proteins.
Delivery: Positive control included.