ContactUsLinkComponent
Oxygen Consumption Rate Assay Kit, Cayman Chemical
Oxygen Consumption Rate Assay Kit, Cayman Chemical
Catalog # 75817-070
CAS Number:  
undefined
Oxygen Consumption Rate Assay Kit, Cayman Chemical
Catalog # 75817-070
Supplier Number:  600800-96
CAS Number:  
Specifications
  • Brand Name:
    CAYM
  • Cat. No.:
    75817-070
  • Contents:
    Kit contains: Probe and Reagents
  • Supplier no.:
    600800-96
Specifications
About this item
For the measurement of oxygen consumption rate in living cells
  • Measure OCR without the need for an oxygen electrode
  • Includes antimycin A, an inhibitor of oxygen consumption, as a control
  • Includes glucose oxidase as a reference for oxygen depletion
The oxygen consumption rate (OCR) of cells is an important indicator of normal cellular function. It is used as a parameter to study mitochondrial function as well as a marker of factors triggering the switch from healthy oxidative phosphorylation to aerobic glycolysis in cancer cells. Oxygen consumption is traditionally measured by a cumbersome oxygen electrode, a specialized piece of equipment that typically yields low sample throughput. A Phosphorescent Oxygen Probe has proven to be useful in measuring oxygen consumption rates in whole cells. Cayman’s cell-based Oxygen Consumption Rate Assay Kit utilizes this newly developed phosphorescent oxygen probe to measure oxygen consumption rate in living cells. Antimycin A, an inhibitor of the mitochondrial electron transport chain, is included to be used as a positive control. Glucose oxidase is also included in the kit to be used as a reference for oxygen depletion. The kit is easy to use and can be easily adapted to high throughput screening for compounds which modulate oxygen consumption rate.