Order Entry
Puerto Rico
ContactUsLinkComponent
β-Hydroxybutyrate (Ketone Body) Assay Kit, Cayman Chemical Company
β-Hydroxybutyrate (Ketone Body) Assay Kit, Cayman Chemical Company
Catalog # 75817-116
CAS Number:  
undefined
β-Hydroxybutyrate (Ketone Body) Assay Kit, Cayman Chemical Company
Catalog # 75817-116
Supplier Number:  700190-96
CAS Number:  

Specifications

  • Brand Name:
    CAYM
  • Cat. No.:
    75817-116
  • Contents:
    Kit contains: Buffer, Standard, Enzyme Solution, Colorimetric Detector, and Plate
  • Supplier no.:
    700190-96

Specifications

About this item

A sensitive tool for measuring β-HB levels in plasma, serum, or urine.

  • Measure β-HB from plasma, serum, urine, cell lysates, and tissue homogenates
  • Assay 40 samples in duplicate
  • Assay Range: 25 – 500 μM
  • Plate-based colorimetric measurement (445 – 455 nm)

β-HB is a 'ketone body' which is produced in the liver, mainly from the oxidation of fatty acids, and is exported to peripheral tissues for use as an energy source. Normally ketosis can indicate that lipid metabolism has been activated and the pathway of lipid degradation is intact. Pathological causes of ketosis include multiple organ failure, diabetes, childhood hypoglycemia, corticosteroid or growth hormone deficiency, intoxication with alcohol or salicylates and several inborn errors of metabolism. Cayman’s β-HB (Ketone Body) Assay provides a simple, reproducible, and sensitive tool for measuring β-HB levels in plasma, serum, or urine in a 96-well plate format with a colorimetric readout at 445 to 455 nm.