Specifications
- Description:Red Handle Scissors, Angled
- Environmentally Preferable:
- Cat. No.:76048-940
- Blade length:100 mm (4")
Specifications
About this item
Red handle scissors with 4 in stainless steel blades. Cuts gauze, tape, clothing and bandages.
The scissors are ideal for first aid kits or as a standalone supply. The compact size allows them to be stored and used on the go.