Order Entry
Puerto Rico
ContactUsLinkComponent
Accessories for UVP Chromato-Vue® Viewing Cabinets, Analytik Jena
Accessories for UVP Chromato-Vue® Viewing Cabinets, Analytik Jena
Catalog # 21474-943
Supplier:  Analytik Jena US
Product available on GSA Advantage®
Small Business Enterprise
Accessories for UVP Chromato-Vue® Viewing Cabinets, Analytik Jena
Catalog # 21474-943
Supplier:  Analytik Jena US
Supplier Number:  34-0007-01
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Description:
    Replacement Tube-C65, 254 nm, 8W
  • Cat. No.:
    21474-943
  • For model:
    C65
  • Supplier no.:
    34-0007-01

Specifications

About this item