DELFIA Assay Buffer is a buffered protein and detergent solution intended for use in DELFIA assays that include lanthanide-labelled proteins
- Harmonized Code US-3822005090
It is optimized to give minimal fluorescence background and non-specific binding in solid phase DELFIA time-resolved fluorescence (TRF) assays. The formulation includes ready-to-use Tris-HCl buffered NaCl solution (pH 7.8) containing 0.05% NaN3, bovine serum albumin (BSA), bovine gamma globulins, Tween 40, diethylenetriaminpentaacetic acid (DTPA) and an inert red dye.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Detection Method:Time-Resolved Fluorescence (TRF),DELFIA TRF
- Storage Temperature:2 to 8 °C
- Brand Name:DELFIA
- Product Type:Reagents