About this item
The DELFIA® Enhancement Solution is an acidic chelating detergent solution intended for use in the quantitative determination of Eu3+/Sm3+ when using a time-resolved fluoroimmunoassay
- Harmonized Code US-3822005090
The solution dissociates Eu3+/Sm3+ from solid phase bound Eu-labeled antibodies or proteins during a time period of a few minutes to form a homogeneous and highly fluorescent micellar chelate solution. The solution allows highly sensitive Eu3+/Sm3+ measurements to be made when using the time-resolved fluorometer. The DELFIA® Enhancement Solution is also used as part of Tb3+/Dy3+ measurement together with DELFIA® Enhancer. Because DELFIA TRF chelates are not intrinsically fluorescent, this solution (an acidic chelating detergent solution) is added to the microplate wells after a wash step to develop the fluorescence signal. This provides highly sensitive Eu and/or Sm measurements. Suitable for use in all multianalyte assays.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Detection Method:Time-Resolved Fluorescence (TRF),DELFIA TRF
- Brand Name:DELFIA
- Product Type:Reagents