Anionic Gold Tracer, Electron Microscopy Sciences
About this item
Anionic tracers are used to visualize positive charged moieties in specimens
- Particle sizes of 0 8, 6, 10, 15 and 25 nm
- Guaranteed shelf life of 18 months
- Supplied in PBS with 15 mM NaN3
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones), cationic tracers bind to polyanionic moieties (membranes, acidic proteins).
10 Options Available Below
Product Details & Documents
Anionic tracers are used to visualize positive charged moieties in specimens
- Particle sizes of 0 8, 6, 10, 15 and 25 nm
- Guaranteed shelf life of 18 months
- Supplied in PBS with 15 mM NaN3
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones), cationic tracers bind to polyanionic moieties (membranes, acidic proteins).
Product Family Options
Product Information
- VolumeSizeAvailabilityPrice
- 100487-896EM Anionic Gold TracerSupplier Number: 25470
- 100487-894EM Anionic Gold TracerSupplier Number: 25469
- 100487-898EM Anionic Gold TracerSupplier Number: 25472
- 100487-900EM Anionic Gold TracerSupplier Number: 25473
- 100487-882EM Anionic Gold TracerSupplier Number: 25460
- 100487-888EM Anionic Gold TracerSupplier Number: 25464
- 100487-886EM Anionic Gold TracerSupplier Number: 25463
- 100487-890EM Anionic Gold TracerSupplier Number: 25466
- 100487-892EM Anionic Gold TracerSupplier Number: 25467
- 100487-884EM Anionic Gold TracerSupplier Number: 25461