Order Entry
Puerto Rico
ContactUsLinkComponent
22 Piece Blood borne Pathogen (BBP) Spill Clean-Up Pack, First Aid Only
22 Piece Blood borne Pathogen (BBP) Spill Clean-Up Pack, First Aid Only
Catalog # 76048-958
Supplier:  First Aid Only
undefined
22 Piece Blood borne Pathogen (BBP) Spill Clean-Up Pack, First Aid Only
Catalog # 76048-958
Supplier:  First Aid Only
Supplier Number:  21-760-001

Specifications

  • Description:
    22 Piece Blood borne Pathogen (BBP) Spill Clean-Up Pack
  • Cat. No.:
    76048-958

Specifications

About this item

All-in-one blood borne pathogen spill clean-up refill pack includes wipes, gloves, disinfectant, scoop, spill powder, and disposable bags.

  • Assortment of disposable tools and personal protective supplies
  • Suitable for the cleanup of blood borne pathogens or other potentially infectious substances

Includes:
Absorbent powder, 2 oz. bag
Scoop/spatula plastic
BZK antiseptic wipes
Nitrile exam gloves, size large
Paper Towels
24×24" biohazard bag
Twist ties
Antimicrobial wipes

Sized to fit into a #10 kit or larger. Also works well as a standalone clean-up kit.