Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant DJ-3 (from E. coli)
Human Recombinant DJ-3 (from E. coli)
Catalog # 103010-650
Supplier:  Anaspec Inc
CAS Number:  
Human Recombinant DJ-3 (from E. coli)
Catalog # 103010-650
Supplier:  Anaspec Inc
Supplier Number:  AS-72219-500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    500 µg
  • Storage Conditions:
    Store at 2-4 °C for immediate use within 1-2 weeks or at –80 °C for up to 12 months. Avoid repeated freeze-thaw cycles.
  • Gene ID:
    NP_001116849
  • Protein Synonyms:
    PARK7|DJ - 1|Human DJ-1|Human PARK7
  • Protein/Peptide Name:
    DJ-3
  • Purity:
    > 95% as determined by SDS-PAGE
  • Molecular Weight:
    19.9 kDa
  • Cat. No.:
    103010-650
  • Supplier no.:
    AS-72219-500

Specifications

About this item

The sequence (Accession # NP_001116849) corresponding to the full length human DJ-1 protein along with N-terminal GST tag was expressed in E. coli. The recombinant DJ-1 protein was purified from bacterial lysate using GST affinity chromatography followed by GST tag cleavage and removal. The molecular weight of the recombinant DJ-1 protein is 19.9 kDa.
DJ-1 is also known as PARK7 (Parkinson disease protein 7). It is a 189-amino acid protein that is ubiquitously expressed in many organs including brain, liver, kidney, pancreas, heart, and others. Although DJ-1 was originally discovered as a novel oncogene product, it was found to play several other roles in biological processes. This includes the regulation of RNA binding activity, fertility, anti-oxidative stress, and is linked to the early onset of Parkinson disease when mutated. Sequence (Accession# NP_001116849) corresponds to the full length human DJ-1 protein and was expressed in E. coli.