Specifications
- Description:ProteoExtract® Transmembrane Protein Extraction Kit
- Size:1 Kit
- Cat. No.:80017-516
- Supplier no.:71772-3
Specifications
About this item
ProteoExtract® Transmembrane Protein Extraction Kit (TM-PEK) uses a novel, detergent-free chemistry for
Each TM-PEK kit provides sufficient reagents to process a total of 40 samples (20 samples using Extraction Buffer 2A at the recommended dilution, and 20 samples using Extraction Buffer 2B at the recommended dilution). Each sample may be comprised of 1–5 x 107 cultured cells or 25–50 mg of tissue.