Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium Citrate Buffers, Sterile, Teknova
Sodium Citrate Buffers, Sterile, Teknova
  100219-862
 :  Teknova
 :  
Sodium Citrate Buffers, Sterile, Teknova
  100219-862
 :  Teknova
 :  S0231
 :  

 

  • Color:
    Colorless
  • Description:
    0.1 M Sodium citrate,sterile
  • Size:
    1 L
  • Storage Temperature:
    Ambient
  • pH buffer type:
    Citrate
  • Cat. No.:
    100219-862
  • pH:
    5.0
  • Supplier no.:
    S0231

 

 

0.1 M sodium citrate, pH adjusted to 5.0 with sodium hydroxide.