ContactUsLinkComponent
CHECKIT® Ozone Test Kits, DPD, Lovibond®
CHECKIT® Ozone Test Kits, DPD, Lovibond®
Catalog # 89212-584
Supplier:  Lovibond
Small Business Enterprise
CAS Number:  
CHECKIT® Ozone Test Kits, DPD, Lovibond®
Catalog # 89212-584
Supplier:  Lovibond
Supplier Number:  147275
Small Business Enterprise
CAS Number:  
Specifications
  • Chemical method:
    DPD (N,N-diethyl-p-phenylenediamine)
  • Description:
    Ozone Test Kit
  • Cat. No.:
    89212-584
  • Range:
    0 - 1.0 mg/L O₃
  • Supplier no.:
    L147275
Specifications
About this item
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.