Order Entry
Puerto Rico
ContactUsLinkComponent
Capto™ S ImpAct Ion Exchange Chromatography Media, Cytiva
Capto™ S ImpAct Ion Exchange Chromatography Media, Cytiva
Supplier:  Cytiva
Certificates
undefined
Capto™ S ImpAct Ion Exchange Chromatography Media, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • pH Range:
    4 - 12
  • Matrix:
    High-flow agarose
  • Ligand:
    -SO3-
  • Storage Temperature:
    4 - 30 °C
  • Binding Capacity:
    > 85 mg BSA/mL medium, > 90 mg lysozyme/mL medium, > 100 mg IgG/ mL medium
  • Total Ionic Capacity:
    37 - 63 umol (H+)/ml

Specifications