About this item
This Ni-NTA Agarose is used for purification of His-tagged proteins by gravity-flow chromatography.
- One-step purification from crude lysate to >95% pure protein
- High binding affinity and high capacity
- Choice of purification under native or denaturing conditions
- Precharged, ready-to-use matrices for any scale of purification
- Automated purification and assay protocols
Ni-NTA Agarose is an affinity chromatography matrix for purifying recombinant proteins carrying a His tag. Histidine residues in the His tag bind to the vacant positions in the coordination sphere of the immobilized nickel ions with high specificity and affinity. Cleared cell lysates are loaded onto the matrices. His-tagged proteins are bound, and other proteins pass through the matrix. After washing, His-tagged proteins are eluted in buffer under native or denaturing conditions.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Max. Pressure:2.8 psi