Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Sirtuin (from E. coli)
Human Recombinant Sirtuin (from E. coli)
Catalog # 103010-584
Supplier:  Anaspec Inc
CAS Number:  
Human Recombinant Sirtuin (from E. coli)
Catalog # 103010-584
Supplier:  Anaspec Inc
Supplier Number:  AS-72187
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    10 µg
  • Storage Conditions:
    –80 °C
  • Gene ID:
    NM_030593
  • Protein Synonyms:
    Human Sirtuin 2|Sirtuin 2 Recombinant|Sirtuin 2|Sirtuin 2 His tag
  • Protein/Peptide Name:
    Sirtuin
  • Molecular Weight:
    35.5 kDa
  • Cat. No.:
    103010-584
  • Supplier no.:
    AS-72187

Specifications

About this item

This human recombinant SIRT2 is in its active form, and can be used for enzyme activity assays. The recombinant human Sirtuin 2 (GenBank Accession #: NM_030593) with 13-319 amino acids and His tag at its C-terminal was expressed in E. coli. The molecular mass of the enzyme is approximately 35.5 kDa on SDS-PAGE.
Sirtuins comprise a unique class of nicotinamide adenine dinucleotide (NAD+)-dependent deacetylases (class III HDACs) targeting multiple protein substrates.
Sirtuin 1 (SIRT1), the human homolog of yeast Sir2 (Silent Information Regulator 2), is the most studied of the seven members of sirtuin family. SIRT1 have been implicated in several important cellular processes, including genomic stability and DNA repair, p53-mediated apoptosis, adipogenesis, and aging.
Substrates for SIRT2 are not limited to histones but also include various transcription factors and co-regulators that modulate metabolic, cell cycle and cell death related pathways. Human SIRT2 is a cytoplasmic protein that increases in abundance during mitosis and regulates major events of cytokinesis.