Order Entry
Puerto Rico
ContactUsLinkComponent
Staphylococcus aureus Recombinant Protein A (from Bacteria)
Staphylococcus aureus Recombinant Protein A (from Bacteria)
Catalog # 103010-688
Supplier:  Anaspec
CAS Number:  
Staphylococcus aureus Recombinant Protein A (from Bacteria)
Catalog # 103010-688
Supplier:  Anaspec
Supplier Number:  AS-72233-5
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Bacteria
  • Species:
    Staphylococcus aureus
  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Gene ID:
    AAB05743.1
  • Protein/Peptide Name:
    Protein A
  • Purity:
    > 95% as determined by SDS-PAGE
  • Molecular Weight:
    ~30 kDa
  • Cat. No.:
    103010-688
  • Supplier no.:
    AS-72233-5

Specifications

About this item

AnaSpec's recombinant Protein A consists of only IgG binding domains and was expressed using a recombinant bacterial expression system. Its apparent molecular weight is approximately 30,000 Da compared to 42,000 Da for the native Protein A that is found in Staphylococcus aureus.
Protein A is a non-glycosylated cell wall protein of Staphylococcus aureus that can bind the Fc part of immunoglobulin molecule of different species with strong affinity (1-4). Protein A consists of five IgG binding domains (A, B, C, D, and E), each approximately 60 amino acids long with no cysteines present and two S. aureus cell membrane binding domains, X and M (3).
Application: Recombinant Protein A can be used for immunoprecipitation, antibodies purification, and assay development.