Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
EndoClear™ Human Recombinant alpha-Synuclein (from <i>E. coli</i>)

EndoClear™ Human Recombinant alpha-Synuclein (from E. coli)

Marketsource
Supplier:   Anaspec
EndoClear™ Human Recombinant alpha-Synuclein (from <i>E. coli</i>)
Supplier:  Anaspec
Marketsource
Certificates
EndoClear™ Human Recombinant alpha-Synuclein (from E. coli)

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Full-length Recombinant human alpha-synuclein (GenBank Accession # NP_000336) was expressed in E. coli., and purified from bacterial lysate using proprietary method. Endotoxin was further removed by proprietary technique.
α-Synuclein is a major component of Lewy bodies in the affected neurons in Parkinson's disease. This protein has a mass of 14.5 kDa (140 amino acids long) and consists of a conserved degenerative amino-terminal domain and an acidic carboxyl-terminal with higher sequence divergence. α-Synuclein is predominantly expressed in brain, specifically in cerebellum, thalamus, neocortex, hippocampus, and striatum regions. Other tissues express α-Synuclein at very low levels. The physiological role of α-synuclein is not yet well understood. However, the presence of imperfect KTKEGV lipid interacting repeats suggests that it may be involved in synaptic vesicle homeostasis.

The human α-synuclein recombinant proteins from AnaSpec are offered with low endotoxin levels of <0.1 EU per μg protein labeled as EndoClear™ products.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 103001-642
  • 103001-646
  • 103001-644
  • Protein/Peptide Name:
  • alpha-Synuclein
  • alpha-Synuclein
  • alpha-Synuclein
  • Protein Synonyms:
  • Synuclein Protein|SNCA|low endotoxin synuclein|alpha-Synuclein|low endotoxin alpha-Synuclein|Human alpha - Synuclein|alpha - Synuclein
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • NP_000336
  • NP_000336
  • NP_000336
  • Endotoxin Level:
  • Low
  • Low
  • Low
  • Endotoxin Content:
  • <0.1EU/µg protein
  • <0.1EU/µg protein
  • <0.1EU/µg protein
  • Purity:
  • > 95% as determined by SDS-PAGE
  • > 95% as determined by SDS-PAGE
  • > 95% as determined by SDS-PAGE
  • Molecular Weight:
  • 14.5 kDa
  • 14.5 kDa
  • 14.5 kDa
  • Storage Conditions:
  • Store at 2-4 °C for immediate use within 1-2 weeks or at –80 °C for up to 12 months.
  • Store at 2-4 °C for immediate use within 1-2 weeks or at –80 °C for up to 12 months.
  • Store at 2-4 °C for immediate use within 1-2 weeks or at –80 °C for up to 12 months.

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 µg
    Log in for availability
    Log in for availability
    Each
    Each - $194.38
    $194.38
    Each
       $194.38

    Supplier Number: AS-55555-100
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103001-642
    Protein/Peptide Name
    alpha-Synuclein
    Protein Synonyms
    Synuclein Protein|SNCA|low endotoxin synuclein|alpha-Synuclein|low endotoxin alpha-Synuclein|Human alpha - Synuclein|alpha - Synuclein
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    NP_000336
    Endotoxin Level
    Low
    Endotoxin Content
    <0.1EU/µg protein
    Purity
    > 95% as determined by SDS-PAGE
    Molecular Weight
    14.5 kDa
    Storage Conditions
    Store at 2-4 °C for immediate use within 1-2 weeks or at –80 °C for up to 12 months.
    Marketsource
  • 500 µg
    Log in for availability
    Log in for availability
    Each
    Each - $584.62
    $584.62
    Each
       $584.62

    Supplier Number: AS-55555-500
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103001-646
    Protein/Peptide Name
    alpha-Synuclein
    Protein Synonyms
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    NP_000336
    Endotoxin Level
    Low
    Endotoxin Content
    <0.1EU/µg protein
    Purity
    > 95% as determined by SDS-PAGE
    Molecular Weight
    14.5 kDa
    Storage Conditions
    Store at 2-4 °C for immediate use within 1-2 weeks or at –80 °C for up to 12 months.
    Marketsource
  • 1000 µg
    Log in for availability
    Log in for availability
    Each
    Each - $973.38
    $973.38
    Each
       $973.38

    Supplier Number: AS-55555-1000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    103001-644
    Protein/Peptide Name
    alpha-Synuclein
    Protein Synonyms
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    NP_000336
    Endotoxin Level
    Low
    Endotoxin Content
    <0.1EU/µg protein
    Purity
    > 95% as determined by SDS-PAGE
    Molecular Weight
    14.5 kDa
    Storage Conditions
    Store at 2-4 °C for immediate use within 1-2 weeks or at –80 °C for up to 12 months.
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.