ContactUsLinkComponent
SP Bel-Art Piezo Electric Gas Lighter, Bel-Art Products, a part of SP
SP Bel-Art Piezo Electric Gas Lighter, Bel-Art Products, a part of SP
Catalog # 490012-186
SP Bel-Art Piezo Electric Gas Lighter, Bel-Art Products, a part of SP
Catalog # 490012-186
Supplier Number:  H15056-0000
Specifications
  • Description:
    Piezo electric gas lighter
  • Length:
    26.5 cm (10⁷⁄₁₆")
  • Cat. No.:
    490012-186
  • Weight:
    3.1 oz.
  • Supplier no.:
    H15056-0000
Specifications
About this item
For lighting Bunsen burners, gas stoves, propane or oxy-acetylene torches safely.
  • Does not require batteries or fuel refills
  • Comfortable plastic pistol-grip with push-button trigger creates an electrical spark (not a flame) at 20 kv with each squeeze