Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Potassium iodide 1 M

Potassium iodide 1 M

Marketsource
Supplier:   Rigaku Reagents
Potassium iodide 1 M
Supplier:  Rigaku Reagents
Marketsource
Certificates
Potassium iodide 1 M
Potassium iodide 1 M

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $86.31
    $86.31
    Each (100ml)
       $86.31

    Supplier Number: 1008202
    Specifications:
    Marketsource
    Marketsource
  • 250 ml
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $136.00
    $136.00
    Each (250ml)
       $136.00

    Supplier Number: 1008203
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.