Order Entry
Puerto Rico
ContactUsLinkComponent
Cadmium(II) chloride 1 M
Cadmium(II) chloride 1 M
Catalog # 101443-260
Supplier:  Rigaku Reagents
CAS Number:  
undefined
Cadmium(II) chloride 1 M
Catalog # 101443-260
Supplier:  Rigaku Reagents
Supplier Number:  1008153
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 ml
  • Cat. No.:
    101443-260
  • Supplier no.:
    EB-1CDCL-100

Specifications

About this item

 :
 :183.32 g/mol
UN:2570
ADR:6.1,III
CAS Number:10108-64-2
Formula:CdCl₂
MDL Number:MFCD00010916