Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:SensoLyte® 520 Cathepsin K Assay Kit *Fluorimetric*
- Size:1 Kit
- Storage Temperature:-20°C
- Cat. No.:103010-546
- No. of wells:96
- No. of assays/kit:100 assays
- Supplier no.:AS-72171
Cathepsin K is the lysosomal cysteine protease involved in bone remodeling and resorption, also having a potential as a drug target in autoimmune diseases and osteoporosis
Cathepsins are a class of globular lysosomal proteases playing a vital role in mammalian cellular turnover. They degrade polypeptides and are distinguished by their substrate specificities.
The cathepsin assay kits offers a series of fluorimetric long wavelength, sensitive SensoLyte® 520 assay kits for enzyme activity determination of Cathepsins G, D, E, B, K, L and S. These kits employ a FRET (Fluorescence Resonance Energy Transfer) peptide substrate that emits in the green range of 520 nm, with excitation at 490 nm. The advantage of using longer wavelength emitting substrates means that autofluorescence from cellular components and test compounds is minimal; thus making the kits more sensitive. Furthermore, the use of HiLyte Fluor™ 488 (except for Cathepsin S, which uses 5-FAM) as the FRET donor (fluorophore) results in a more stable fluorescent signal, since HiLyte Fluor™ 488 is resistant to acidic pH. A listing of the SensoLyte® Cathepsin Kits providing conventional FRET (Mca/Dnp) or fluorogenic substrates for cathepsins are also available.
Kits detect specific Cathepsin activities; provides ample reagents to perform 100 assays in 96-well format; minimal hands-on time required; detailed protocol and references provided.
SensoLyte® 520 Cathepsin K Activity Assay Kit uses a QXL® 520/HiLyte™ Fluor 488 FRET peptide substrate for the measurement of enzyme activity. In the intact FRET peptide, the fluorescence of HiLyte™ Fluor 488 is quenched by QXL® 520. Upon cleavage of the FRET peptide by the active enzyme, the increase of fluorescence can be continuously monitored at excitation/emission = 490/520 nm. The fluorescent signal from HiLyte™ Fluor 488 is stable at low pH, which is optimal for cathepsin activity. The kit can be used to detect the activity of Cathepsin K enzyme in biological samples and purified enzyme preparations.