Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® FDP Protein Phosphatase Assay Kit Fluorimetric, AnaSpec, Inc.
SensoLyte® FDP Protein Phosphatase Assay Kit Fluorimetric, AnaSpec, Inc.
Catalog # 103010-108
Supplier:  Anaspec
CAS Number:  
SensoLyte® FDP Protein Phosphatase Assay Kit Fluorimetric, AnaSpec, Inc.
Catalog # 103010-108
Supplier:  Anaspec
Supplier Number:  AS-71100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    SensoLyte® FDP Protein Phosphatase Assay Kit *Fluorimetric*
  • Size:
    1 Kit
  • Storage Temperature:
    - 20 °C
  • Cat. No.:
    103010-108
  • No. of wells:
    96
  • No. of assays/kit:
    500 assays (96 well plate) or 1250 assays (384 well plate)
  • Supplier no.:
    AS-71100

Specifications

About this item

FDP is a highly sensitive fluorogenic substrate for most phosphatases

Protein phosphatases have received great attention as drug-screening targets. SensoLyte® Protein Phosphatase Assay Kits uses specific substrates to quantify specific protein phosphatase activities such as MFP, pNPP FDP or acid phosphatases. The kit can be used for characterizing kinetics of enzyme reaction and high throughput screening of protein phosphatase inhibitors/inducers.

Optimized to detect protein phosphatase activity; compatible with HTS; provides enough reagents to perform 500 assays in a 96-well format; minimal hands-on time; detailed protocol and references provided.

The phosphatase-induced hydrolysis of FDP yields an intense fluorescent product that has excitation wavelength around 490 nm, and emission maximum around 520 nm. The fluorescence of FDP hydrolyzed product is strengthened when pH > 7.0. For some protein phosphatase requiring slightly acidic buffer for reaction, a pH-adjusting stop solution should be added before reading the fluorescence signal. The kit can be used for characterizing kinetics of enzyme reaction and high throughput screening of protein phosphatase inducers and inhibitors.