Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® Luminescent Alkaline Phosphatase Assay Kit Luminometric
SensoLyte® Luminescent Alkaline Phosphatase Assay Kit Luminometric
Catalog # 103010-452
Supplier:  Anaspec
CAS Number:  
SensoLyte® Luminescent Alkaline Phosphatase Assay Kit Luminometric
Catalog # 103010-452
Supplier:  Anaspec
Supplier Number:  AS-72122
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    SensoLyte® Luminescent Alkaline Phosphatase Assay Kit *Luminometric*
  • Size:
    1 Kit
  • Storage Temperature:
    4 °C
  • Cat. No.:
    103010-452
  • No. of wells:
    96
  • No. of assays/kit:
    500 assays
  • Supplier no.:
    AS-72122

Specifications

About this item

SensoLyte® Luminescent Alkaline Phosphatase Assay Kit provides highly sensitive chemiluminescent substrate to quantify alkaline phosphatase activity in solutions, in cell extracts, in live cells.

Phosphatases are important classes of enzymes involved in cellular signal transduction, cell metabolism and other biological activities. Changes in alkaline phosphatase level and activity are involved in a variety of physiological and pathological events, such as bone development, bone-related diseases, gestation related diseases, inflammatory bowel disease, post-parathyroidectomy stage and drug toxicity. Phosphatases have attracted considerable interest as: Enzyme markers, Drug screening targets, Conjugating agents and Reporter for gene expression studies.

Features: Kits include all essential components (chromogenic/fluorogenic/luminometry substrates) and sample reagents needed to perform 500 assays using 96-well plate format.
Optimized to detect specific alkaline phosphatase activity; compatible for HTS; detailed protocol and references provided.