Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® pNPP Alkaline Phosphatase Assay Kit Colorimetric, AnaSpec, Inc.
SensoLyte® pNPP Alkaline Phosphatase Assay Kit Colorimetric, AnaSpec, Inc.
Catalog # 103010-494
Supplier:  Anaspec
CAS Number:  
SensoLyte® pNPP Alkaline Phosphatase Assay Kit Colorimetric, AnaSpec, Inc.
Catalog # 103010-494
Supplier:  Anaspec
Supplier Number:  AS-72146
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    SensoLyte® pNPP Alkaline Phosphatase Assay Kit *Colorimetric*
  • Size:
    1 Kit
  • Storage Temperature:
    4 °C
  • Cat. No.:
    103010-494
  • No. of wells:
    96
  • No. of assays/kit:
    500 assays
  • Supplier no.:
    AS-72146

Specifications

About this item

Alkaline phosphatase is a popular enzyme conjugated with secondary antibody for ELISA

  • Features: Kits include all essential components (chromogenic/fluorogenic/luminometry substrates) and sample reagents needed to perform 500 assays using 96-well plate format.
  • Optimized to detect specific alkaline phosphatase activity; compatible for HTS; detailed protocol and references provided.

Phosphatases are important classes of enzymes involved in cellular signal transduction, cell metabolism and other biological activities. Changes in alkaline phosphatase level and activity are involved in a variety of physiological and pathological events, such as bone development, bone-related diseases, gestation related diseases, inflammatory bowel disease, post-parathyroidectomy stage and drug toxicity. Phosphatases have attracted considerable interest as: Enzyme markers, Drug screening targets, Conjugating agents and Reporter for gene expression studies.

p-Nitrophenyl phosphate (pNPP) is proven to be an effective chromogenic substrate for alkaline phosphate. This SensoLyte® kit can be used to detect alkaline phosphatase activity in biological samples and alkaline phosphatase-based ELISA.

Ordering information: Kits include all essential components (chromogenic/fluorogenic/luminometry substrates) and sample reagents needed to perform 500 assays using 96-well plate format.