Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® pNPP Secreted Alkaline Phosphatase Reporter Gene Assay Kit Colorimetric, AnaSpec, Inc.
SensoLyte® pNPP Secreted Alkaline Phosphatase Reporter Gene Assay Kit Colorimetric, AnaSpec, Inc.
Catalog # 103010-490
Supplier:  Anaspec
CAS Number:  
SensoLyte® pNPP Secreted Alkaline Phosphatase Reporter Gene Assay Kit Colorimetric, AnaSpec, Inc.
Catalog # 103010-490
Supplier:  Anaspec
Supplier Number:  AS-72144
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    SensoLyte® pNPP Secreted Alkaline Phosphatase Reporter Gene Assay Kit *Colorimetric*
  • Size:
    1 Kit
  • Storage Temperature:
    -20°C
  • Cat. No.:
    103010-490
  • No. of wells:
    96
  • No. of assays/kit:
    500 assays
  • Supplier no.:
    AS-72144

Specifications

About this item

The secreted alkaline phosphatase (SEAP) is widely used as reporter gene to analyze gene expression including kinetic analysis over time in cell culture or animals owing to its unique ability to secrete into culture medium and serum

  • Features: Kits include all essential components (chromogenic/fluorogenic/luminometry substrates) and sample reagents needed to perform 500 assays using 96-well plate format.
  • Optimized to detect specific alkaline phosphatase activity; compatible for HTS; detailed protocol and references provided.

Phosphatases are important classes of enzymes involved in cellular signal transduction, cell metabolism and other biological activities. Changes in alkaline phosphatase level and activity are involved in a variety of physiological and pathological events, such as bone development, bone-related diseases, gestation related diseases, inflammatory bowel disease, post-parathyroidectomy stage and drug toxicity. Phosphatases have attracted considerable interest as: Enzyme markers, Drug screening targets, Conjugating agents and Reporter for gene expression studies.

SensoLyte® pNPP Secreted Alkaline Phosphatase Reporter Gene Assay Kit provides a sensitive colorimetric assay of placental alkaline phosphatase for both secreted and membrane-bound form of enzyme

Ordering information: Kits include all essential components (chromogenic/fluorogenic/luminometry substrates) and sample reagents needed to perform 500 assays using 96-well plate format.