Orders LinkContactUsLinkComponent
Demonstration Electroscope
Demonstration Electroscope
Demonstration Electroscope
Catalog # 470230-320
Supplier:   Wards
Supplier #:  PH0917A
Demonstration Electroscope
Catalog # 470230-320
Supplier:  Wards
Demonstration Electroscope
Demonstration Electroscope
Demonstration Electroscope
Price
$52.40
Log in for availability
Each
Product Details & Documents
Demonstrates static potentials.
  • 150 mm diameter ring
For the demonstration of static potentials. Available with charging ball plus a set of condenser plates with insulated rod. Ring diameter 150 mm.
Specifications
  • Description:
    Demonstration Electroscope